ENA429361000 Enamine

2-(4-methylphenyl)-4H,5H,6H,7H-[1,3]thiazolo[5,4-c]pyridine hydrobromide

  • MDL number MFCD18071281
  • Cas Number : 1328486-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191447

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191447
Availability details for
Quantity
  • Availability
  • Add to Cart