ENA429361009 Enamine

1H,2H,3H,9H-pyrrolo[2,1-b]quinazoline hydrochloride

  • MDL number MFCD00155365
  • Cas Number : 61939-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191457
Availability details for
Quantity
  • Availability
  • Add to Cart