ENA429361016 Enamine

3-amino-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one hydrochloride

  • MDL number MFCD04035624
  • Cas Number : 148502-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191464
Availability details for
Quantity
  • Availability
  • Add to Cart