ENA429361037 Enamine

3-(2-aminoethyl)-5-fluoro-2,3-dihydro-1H-indol-2-one hydrochloride

  • MDL number MFCD11983043
  • Cas Number : 1245568-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191486
Availability details for
Quantity
  • Availability
  • Add to Cart