ENA429361045 Enamine

1-(3-aminophenyl)piperidine-2,6-dione

  • MDL number MFCD10694932
  • Cas Number : 1152523-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191497
Availability details for
Quantity
  • Availability
  • Add to Cart