ENA429361047 Enamine

1-methyl-3-(4,5,6,7-tetrahydro-1-benzothiophen-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD11105303
  • Cas Number : 1152708-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189745
Availability details for
Quantity
  • Availability
  • Add to Cart