ENA429361053 Enamine

tert-butyl 5-chloropyrazine-2-carboxylate

  • MDL number MFCD18070852
  • Cas Number : 169335-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191547
Availability details for
Quantity
  • Availability
  • Add to Cart