ENA429361057 Enamine

ethoxy(ethylsulfanyl)methanethione

  • MDL number MFCD13194830
  • Cas Number : 623-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191556

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191556
Availability details for
Quantity
  • Availability
  • Add to Cart