ENA429361064 Enamine

2-(aminomethyl)pyridin-3-ol

  • MDL number MFCD01217253
  • Cas Number : 194665-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191576
Availability details for
Quantity
  • Availability
  • Add to Cart