ENA429361065 Enamine

4-(2-methoxypropoxy)cyclohexane-1-carboxylic acid

  • MDL number MFCD28125188
  • Cas Number : 1601118-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191578
Availability details for
Quantity
  • Availability
  • Add to Cart