ENA429361069 Enamine

2-[3-(propan-2-yl)pyrrolidin-1-yl]ethan-1-amine dihydrochloride

  • MDL number MFCD28118859
  • Cas Number : 1803580-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189778
Availability details for
Quantity
  • Availability
  • Add to Cart