ENA429361071 Enamine

2-(2-nitrophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD07371521
  • Cas Number : 861337-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-163
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191624
Availability details for
Quantity
  • Availability
  • Add to Cart