ENA429361072 Enamine

3-[5-(difluoromethyl)-1,2,4-oxadiazol-3-yl]aniline

  • MDL number MFCD12191351
  • Cas Number : 1155522-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191626
Availability details for
Quantity
  • Availability
  • Add to Cart