ENA429361073 Enamine

4-bromo-3-ethyl-1,2-oxazol-5-amine

  • MDL number MFCD20702410
  • Cas Number : 166964-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191627
Availability details for
Quantity
  • Availability
  • Add to Cart