ENA429361075 Enamine

[5-(trifluoromethyl)-1,2-oxazol-3-yl]methanol

  • MDL number MFCD18818572
  • Cas Number : 172031-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191629

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191629
Availability details for
Quantity
  • Availability
  • Add to Cart