ENA429361076 Enamine

1-cyclohexylimidazolidin-2-one

  • MDL number MFCD23696280
  • Cas Number : 37732-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191630

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191630
Availability details for
Quantity
  • Availability
  • Add to Cart