ENA429361080 Enamine

3-(piperidin-1-yl)cyclobutan-1-amine

  • MDL number MFCD18381813
  • Cas Number : 878155-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191646

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191646
Availability details for
Quantity
  • Availability
  • Add to Cart