ENA429361084 Enamine

2-methyl-1-(piperidin-4-yl)-1H-1,3-benzodiazole

  • MDL number MFCD14552074
  • Cas Number : 79098-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191670
Availability details for
Quantity
  • Availability
  • Add to Cart