ENA429361088 Enamine

methyl 1-sulfanylcyclohexane-1-carboxylate

  • MDL number MFCD19689209
  • Cas Number : 1378832-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191688

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191688
Availability details for
Quantity
  • Availability
  • Add to Cart