ENA429361100 Enamine

3-cyclopentylazetidine hydrochloride

  • MDL number MFCD22683337
  • Cas Number : 1803607-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191724
Availability details for
Quantity
  • Availability
  • Add to Cart