ENA429361104 Enamine

ethyl 2-cyano-3-(quinolin-2-yl)propanoate

  • MDL number MFCD06211099
  • Cas Number : 773086-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191733
Availability details for
Quantity
  • Availability
  • Add to Cart