ENA429361105 Enamine

2-(5-methyl-1,3-oxazol-2-yl)aniline

  • MDL number MFCD14657133
  • Cas Number : 52829-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189799
Availability details for
Quantity
  • Availability
  • Add to Cart