ENA429361109 Enamine

4-methanesulfonyl-1,2-dimethylbenzene

  • MDL number MFCD03789178
  • Cas Number : 38452-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191759

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191759
Availability details for
Quantity
  • Availability
  • Add to Cart