ENA429361110 Enamine

1-methanesulfonylcyclobutane-1-carboxylic acid

  • MDL number MFCD16766464
  • Cas Number : 1250528-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191771
Availability details for
Quantity
  • Availability
  • Add to Cart