ENA429361111 Enamine

2,6-difluoro-3-iodobenzaldehyde

  • MDL number MFCD11845973
  • Cas Number : 1160573-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191772
Availability details for
Quantity
  • Availability
  • Add to Cart