ENA429361113 Enamine

1-(difluoromethyl)-2-oxo-1,2-dihydropyridine-3-carbaldehyde

  • MDL number MFCD18791578
  • Cas Number : 1263178-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191778

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191778
Availability details for
Quantity
  • Availability
  • Add to Cart