ENA429361115 Enamine

3-[(3-chloropropyl)sulfanyl]propanoic acid

  • MDL number MFCD20623125
  • Cas Number : 16496-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191813

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191813
Availability details for
Quantity
  • Availability
  • Add to Cart