ENA429361117 Enamine

ethyl 2-cyano-2,2-difluoroacetate

  • MDL number MFCD25989269
  • Cas Number : 219866-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191821
Availability details for
Quantity
  • Availability
  • Add to Cart