ENA429361120 Enamine

tert-butyl N-(2-bromo-6-fluorophenyl)carbamate

  • MDL number MFCD24470774
  • Cas Number : 1057720-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191826

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191826
Availability details for
Quantity
  • Availability
  • Add to Cart