ENA429361123 Enamine

1-(2-methoxyethyl)-1H-indole-5-carboxylic acid

  • MDL number MFCD18873289
  • Cas Number : 1304787-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191867
Availability details for
Quantity
  • Availability
  • Add to Cart