ENA429361124 Enamine

4-methyl-2-[(2-methylpropyl)amino]-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD08165214
  • Cas Number : 927983-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191868
Availability details for
Quantity
  • Availability
  • Add to Cart