ENA429361125 Enamine

2-[2-(2,3-dichlorophenyl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD11179463
  • Cas Number : 1094293-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191869
Availability details for
Quantity
  • Availability
  • Add to Cart