ENA429361127 Enamine

5-amino-1-(2-chloro-5-fluorophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD17430370
  • Cas Number : 1232770-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191871
Availability details for
Quantity
  • Availability
  • Add to Cart