ENA429361128 Enamine

1-(2-methoxyethyl)-1H-indole-6-carboxylic acid

  • MDL number MFCD18873302
  • Cas Number : 1304787-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191872
Availability details for
Quantity
  • Availability
  • Add to Cart