ENA429361130 Enamine

4-methyl-2-(1H-pyrrol-1-yl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD11986883
  • Cas Number : 1105192-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191874
Availability details for
Quantity
  • Availability
  • Add to Cart