ENA429361132 Enamine

2-(cyclopentylamino)-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD08165217
  • Cas Number : 927983-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-191876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191876
Availability details for
Quantity
  • Availability
  • Add to Cart