ENA429361135 Enamine

1-(propan-2-yl)-1H-indole-5-carboxylic acid

  • MDL number MFCD15200971
  • Cas Number : 178053-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191879

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191879
Availability details for
Quantity
  • Availability
  • Add to Cart