ENA429361137 Enamine

2-[1-(2-methoxyethyl)-1H-indol-3-yl]acetic acid

  • MDL number MFCD11155395
  • Cas Number : 1021141-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191881
Availability details for
Quantity
  • Availability
  • Add to Cart