ENA429361139 Enamine

5-oxo-5-(3-oxopiperazin-1-yl)pentanoic acid

  • MDL number MFCD09045202
  • Cas Number : 926237-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191883

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191883
Availability details for
Quantity
  • Availability
  • Add to Cart