ENA429361142 Enamine

2-(4-methyl-1H-indol-1-yl)acetic acid

  • MDL number MFCD11212267
  • Cas Number : 1094641-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191886
Availability details for
Quantity
  • Availability
  • Add to Cart