ENA429361143 Enamine

1-(propan-2-yl)-1H-indole-6-carboxylic acid

  • MDL number MFCD11043021
  • Cas Number : 1030423-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191887
Availability details for
Quantity
  • Availability
  • Add to Cart