ENA429361151 Enamine

3,4-dimethyl-2-(1H-1,2,3,4-tetrazol-1-yl)benzoic acid

  • MDL number MFCD16090152
  • Cas Number : 1219547-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191895

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191895
Availability details for
Quantity
  • Availability
  • Add to Cart