ENA429361154 Enamine

(2S)-2-{1,3-dioxo-1H,2H,3H-pyrrolo[3,4-c]pyridin-2-yl}propanoic acid

  • MDL number MFCD11501573
  • Cas Number : 1212191-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191898

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191898
Availability details for
Quantity
  • Availability
  • Add to Cart