ENA429361157 Enamine

2-(4-acetamido-1H-indol-1-yl)acetic acid

  • MDL number MFCD26523113
  • Cas Number : 1573548-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191901
Availability details for
Quantity
  • Availability
  • Add to Cart