ENA429361163 Enamine

3-(7-methoxy-1H-indol-1-yl)propanoic acid

  • MDL number MFCD16090150
  • Cas Number : 1219587-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191907
Availability details for
Quantity
  • Availability
  • Add to Cart