ENA429361165 Enamine

1-(propan-2-yl)-1H-indole-4-carboxylic acid

  • MDL number MFCD15200972
  • Cas Number : 1030423-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191909
Availability details for
Quantity
  • Availability
  • Add to Cart