ENA429361166 Enamine

3-[(1H-indol-2-yl)formamido]propanoic acid

  • MDL number MFCD11131662
  • Cas Number : 68724-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191910
Availability details for
Quantity
  • Availability
  • Add to Cart