ENA429361169 Enamine

1-oxo-2-(propan-2-yl)-1,2-dihydroisoquinoline-4-carboxylic acid

  • MDL number MFCD20535516
  • Cas Number : 1352524-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191913

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191913
Availability details for
Quantity
  • Availability
  • Add to Cart