ENA429361178 Enamine

3-(5-bromothiophen-2-yl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD03030193
  • Cas Number : 1038558-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191922

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191922
Availability details for
Quantity
  • Availability
  • Add to Cart