ENA429361179 Enamine

3-(4-chlorophenyl)-3-(1H-pyrrol-1-yl)propanoic acid

  • MDL number MFCD04060717
  • Cas Number : 404869-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191923

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191923
Availability details for
Quantity
  • Availability
  • Add to Cart